Merck
  • Method of preparing a decellularized porcine tendon using tributyl phosphate.
Method of preparing a decellularized porcine tendon using tributyl phosphate.
Journal of biomedical materials research. Part B, Applied biomaterials (2011-01-07)
C R Deeken, A K White, S L Bachman, B J Ramshaw, D S Cleveland, T S Loy, S A Grant
ABSTRACT
Extracellular matrix (ECM) materials are currently utilized for soft tissue repair applications such as vascular grafts, tendon reconstruction, and hernia repair. These materials are derived from tissues such as human dermis and porcine small intestine submucosa, which must be rendered acellular to prevent disease transmission and decrease the risk of an immune response. The ideal decellularization technique removes cells and cellular remnants, but leaves the original collagen architecture intact. The tissue utilized in this study was the central tendon of the porcine diaphragm, which had not been previously investigated for soft tissue repair. Several treatments were investigated during this study including peracetic acid, TritonX-100, sodium dodecyl sulfate, and tri(n-butyl) phosphate (TnBP). Of the decellularization treatments investigated, only 1% TnBP was effective in removing cell nuclei while leaving the structure and composition of the tissue intact. Overall, the resulting acellular tissue scaffold retained the ECM composition, strength, resistance to enzymatic degradation, and biocompatibility of the original tissue, making 1% TnBP an acceptable decellularization treatment for porcine diaphragm tendon.
Sigma-Aldrich
Tributyl phosphate, 97%
Sigma-Aldrich
Tributyl phosphate, puriss., ≥99.0% (GC)
Sigma-Aldrich
Tributyl phosphate, ≥99%
Supelco
Tributyl phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tributyl phosphate, for extraction analysis, ≥99.0% (GC)
Tri-n-butyl phosphate, European Pharmacopoeia (EP) Reference Standard