Merck
  • Determination of dexamethasone and dexamethasone sodium phosphate in human plasma and cochlear perilymph by liquid chromatography/tandem mass spectrometry.
Determination of dexamethasone and dexamethasone sodium phosphate in human plasma and cochlear perilymph by liquid chromatography/tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-11-30)
Mei Zhang, Grant A Moore, Berit P Jensen, Evan J Begg, Philip A Bird
ABSTRACT
A rapid, simple and sensitive liquid chromatography/tandem mass spectrometry (LC-MS/MS) assay was developed for the determination of dexamethasone (Dex) and dexamethasone sodium phosphate (Dex SP) in plasma and human cochlear perilymph. After proteins were precipitated with a mixture of acetonitrile and methanol, Dex, Dex SP and flumethasone, the internal standard, were resolved on a C18 column using gradient elution of 5 mM ammonium acetate and methanol. The three compounds were detected using electrospray ionisation in the positive mode. Standard curves were linear over the concentration range 0.5-500 μg/L (r>0.99), bias was <±10%, intra- and inter-day coefficients of variation (imprecision) were <10%, and the limit of quantification was 0.5 μg/L for both Dex and Dex SP. The assay has been used successfully in a clinical pharmacokinetics study of Dex and Dex SP in cochlear perilymph and plasma.
Sigma-Aldrich
Flumethasone