Merck
Layer-by-layer quantum dot constructs using self-assembly methods.
Langmuir : the ACS journal of surfaces and colloids (2010-10-13)
Sakandar Rauf, Andrew Glidle, Jonathan M Cooper
ABSTRACT
We describe the creation of CdSe/ZnS quantum dot assemblies using layer-by-layer construction strategies, using self-assembly. In the first approach, a dithiol linker was used to make multilayers of CdSe/ZnS quantum dots, while in the second biotin- and streptavidin-conjugated CdSe/ZnS quantum dots were used to make multilayer constructs. Both the chemical bonding nature and fluorescence spectroscopic properties of quantum dot films were characterized using X-ray photoelectron spectroscopy (XPS) and fluorescence spectroscopy.
Product Description
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%