Merck
  • Synthesis and electrochemical studies of charge-transfer complexes of thiazolidine-2,4-dione with sigma and pi acceptors.
Synthesis and electrochemical studies of charge-transfer complexes of thiazolidine-2,4-dione with sigma and pi acceptors.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-01-19)
Prashant Singh, Pradeep Kumar, Anju Katyal, Rashmi Kalra, Sujata K Dass, Satya Prakash, Ramesh Chandra
ABSTRACT
In the present work, we report the synthesis and characterization of novel charge-transfer complexes of thiazolidine-2,4-dione (TZD) with sigma acceptor (iodine) and pi acceptors (chloranil, dichlorodicyanoquinone, picric acid and duraquinone). We also evaluated their thermal and electrochemical properties and we conclude that these complexes are frequency dependent. Charge-transfer complex between thiazolidine-2,4-dione and iodine give best conductivity. In conclusion, complex with sigma acceptors are more conducting than with pi acceptors.
Product Description
Supelco
Chloranil, PESTANAL®, analytical standard
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%