Merck
  • The potential dermal irritating effect of residual (meth)acrylic monomers in pressure sensitive adhesive tapes.
The potential dermal irritating effect of residual (meth)acrylic monomers in pressure sensitive adhesive tapes.
Drug and chemical toxicology (2009-12-17)
Fumio Tokumura, Tetsuya Matsui, Yasuko Suzuki, Masashi Sado, Masaharu Taniguchi, Ichiro Kobayashi, Masashi Kamiyama, Shin Suda, Atsushi Nakamura, Yuhiro Yamazaki, Akira Yamori, Ryosuke Igarashi, Jun Kawai, Keiji Oka
ABSTRACT
It is generally thought that residual unpolymerized (meth)acrylic monomers commonly found in pressure sensitive adhesive tapes for medical use may cause dermal irritation, but a systematic study has never been carried out. Therefore, we assessed the potential dermal irritating effect of residual (meth)acrylic monomers. We studied seven acrylic monomers, acrylic acid (AA), methyl acrylate (MA), ethyl acrylate (EA), n-butyl acrylate (n-BA), n-hexyl acrylate (n-HA), 2-ethylhexyl acrylate (2-EHA) and 2-hydroxyethyl acrylate (HEA), as well as three methacrylic monomers, methacrylic acid (MAA), methyl methacrylate (MMA) and 2-hydroxyethyl methacrylate (2-HEMA). We first examined their cytotoxic effect on a cultured dermis model using the MTT method to determine their EC(50) and then performed a primary irritation test in rabbits using the monomers at three different concentrations (i.e., EC(50) , one-tenth EC(50) and 10 times EC(50)). Marked variations were found in cytotoxic and dermal irritating activities among the (meth)acrylic monomers tested. HEA exhibited the most potent dermal irritation having the lowest erythema dose (the concentration which gives a primary dermal irritation index of 1.00) of 460 ppm. But the other monomers exhibited less potent dermal irritation (lowest erythema doses > or =1000 ppm). For the monomers, significant correlation was found between cytotoxic activity and in vivo dermal irritating activity. Our results show that residual unpolymerized (meth)acrylic monomers in adhesive tapes are unlikely to induce skin irritation except for HEA. This study also suggests that cultured skin models are extremely useful as a screening method for chemical substances that could potentially cause dermal irritating activity.
Product Description
Supelco
2-Ethylhexyl acrylate, analytical standard
Supelco
Methyl acrylate, analytical standard
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Ethylhexyl acrylate, 98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer
Sigma-Aldrich
2-Hydroxyethyl acrylate, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor