Merck
  • Separation and preconcentration of trace amounts of aluminum ions in surface water samples using different analytical techniques.
Separation and preconcentration of trace amounts of aluminum ions in surface water samples using different analytical techniques.
Talanta (2009-09-29)
Sumaira Khan, Tasneem G Kazi, Jameel A Baig, Nida F Kolachi, Hassan I Afridi, Abdul Q Shah, Ghulam A Kandhro, Sham Kumar
ABSTRACT
A separation/preconcentration of aluminum (III) (Al(3+)) has been developed to overcome the problem of high matrix species, which may interfere with the determination of trace quantity of Al(3+) in natural water samples. The separation of Al(3+) in water samples was carried out from interfering cations by complexing them with 2-methyle 8-hyroxyquinoline (quinaldine) on activated silica. Whereas the separated trace amounts of Al(3+) was preconcentrated by cloud point extraction (CPE), as prior step to its determination by spectrofluorimetry (SPF) and flame atomic absorption spectrometry (FAAS). The Al(3+) react with 8-hydroxyquinoline (oxine) and then entrapped in non-ionic surfactant Triton X-114. The main factors affecting CPE efficiency, such as pH of sample solution, concentration of oxine and Triton X-114, equilibration temperature and time period for shaking were investigated in detail. The validity of separation/preconcentration of Al(3+) was checked by certified reference material of water (SRM-1643e). After optimization of the complexation and extraction conditions, a preconcentration factor of 20 was obtained for Al(3+) in 10 mL of natural water samples. The relative standard deviation for 6 replicates containing 100 microg L(-1) of Al(3+) was 5.41 and 4.53% for SPF and FAAS, respectively. The proposed method has been applied for determination of trace amount of Al(3+) in natural water samples with satisfactory results.
Product Description
Supelco
Quinaldine, analytical standard, ≥90% (GC)
Sigma-Aldrich
Quinaldine, ≥95.0% (GC)