Merck
  • Early calcium increase triggers the formation of olfactory long-term memory in honeybees.
Early calcium increase triggers the formation of olfactory long-term memory in honeybees.
BMC biology (2009-06-18)
Emmanuel Perisse, Valérie Raymond-Delpech, Isabelle Néant, Yukihisa Matsumoto, Catherine Leclerc, Marc Moreau, Jean-Christophe Sandoz
ABSTRACT
Synaptic plasticity associated with an important wave of gene transcription and protein synthesis underlies long-term memory processes. Calcium (Ca2+) plays an important role in a variety of neuronal functions and indirect evidence suggests that it may be involved in synaptic plasticity and in the regulation of gene expression correlated to long-term memory formation. The aim of this study was to determine whether Ca2+ is necessary and sufficient for inducing long-term memory formation. A suitable model to address this question is the Pavlovian appetitive conditioning of the proboscis extension reflex in the honeybee Apis mellifera, in which animals learn to associate an odor with a sucrose reward. By modulating the intracellular Ca2+ concentration ([Ca2+]i) in the brain, we show that: (i) blocking [Ca2+]i increase during multiple-trial conditioning selectively impairs long-term memory performance; (ii) conversely, increasing [Ca2+]i during single-trial conditioning triggers long-term memory formation; and finally, (iii) as was the case for long-term memory produced by multiple-trial conditioning, enhancement of long-term memory performance induced by a [Ca2+]i increase depends on de novo protein synthesis. Altogether our data suggest that during olfactory conditioning Ca2+ is both a necessary and a sufficient signal for the formation of protein-dependent long-term memory. Ca2+ therefore appears to act as a switch between short- and long-term storage of learned information.
Sigma-Aldrich
1-Nonanol, 98%
Sigma-Aldrich
1-Nonanol, purum, ≥98.0% (GC)
Sigma-Aldrich
Nonyl alcohol, ≥98%, FCC