Merck
Synthesis of zeolite Li-ABW from fly ash by fusion method.
Journal of hazardous materials (2009-06-06)
Z T Yao, M S Xia, Y Ye, L Zhang
ABSTRACT
The zeolite Li-ABW was synthesized by fusion method using fly ash as raw material. It comprised alkaline fusion followed by hydrothermal treatment in LiOH x H2O medium. Crystallinity of zeolite as high as 97.8% was attained under the following conditions: LiOH x H2O concentration, 3M; hydrothermal temperature, 180 degrees C; the corresponding aging time, 12 h. The content of Li-ABW increased at the expense of lithium aluminum silicate or quartz with an increase of LiOH x H2O concentration. With increasing hydrothermal temperature and aging time, the soluble species re-crystallized and crystalline phase transformation between different zeolites was observed. Scanning electron microscopy (SEM) observation revealed that the obtained zeolite Li-ABW was a rod-like crystal. The pore size distribution curve indicated the presence of mesopores.
Sigma-Aldrich
Lithium hydroxide monohydrate, BioXtra, 98.5-101.5% (titration)
Sigma-Aldrich
Lithium hydroxide monohydrate, 99.95% trace metals basis
Sigma-Aldrich
Lithium hydroxide monohydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Lithium hydroxide monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithium hydroxide, reagent grade, 98%
Sigma-Aldrich
Lithium hydroxide, powder, reagent grade, ≥98%