Merck
  • Diethylzinc-mediated one-step stereoselective synthesis of alpha-fluoroacrylates from aldehydes and ketones. Two different pathways depending on the carbonyl partner.
Diethylzinc-mediated one-step stereoselective synthesis of alpha-fluoroacrylates from aldehydes and ketones. Two different pathways depending on the carbonyl partner.
The Journal of organic chemistry (2009-05-06)
G Lemonnier, L Zoute, G Dupas, J-C Quirion, P Jubault
ABSTRACT
A efficient methodology allowing the one-pot stereoselective synthesis of alpha-fluoroacrylates, based on the addition of ethyl dibromofluoroacetate to a carbonyl derivative using diethylzinc as organometallic mediator, is described. Two different pathways have been identified depending on the involved carbonyl partner. In the case of aldehydes, an E2-type mechanism has been identified, whereas ketones go through an E1cb-type mechanism.
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis