Merck
  • Layered material gamma-ZrP supported platinum catalyst for liquid-phase reaction: a highly active and selective catalyst for hydrogenation of the nitro group in para-chloronitrobenzene.
Layered material gamma-ZrP supported platinum catalyst for liquid-phase reaction: a highly active and selective catalyst for hydrogenation of the nitro group in para-chloronitrobenzene.
Chemical communications (Cambridge, England) (2008-06-10)
Fang Wang, Junhua Liu, Xianlun Xu
ABSTRACT
Chemoselective hydrogenation of para-chloronitrobenzene without any dechlorination over Pt/gamma-ZrP catalyst proceeds effectively, which provides a clean and convenient approach to produce useful para-chloroaniline with excellent activity and selectivity.
Sigma-Aldrich
4-Chloroaniline, 98%
Sigma-Aldrich
4-Chloroaniline, purified by sublimation, ≥99%
Supelco
4-Chloroaniline, PESTANAL®, analytical standard
Supelco
p-Chloroaniline, Pharmaceutical Secondary Standard; Certified Reference Material