Merck
  • Adenylate cyclase activity in the families of alcoholics is controlled by a single major gene.
Adenylate cyclase activity in the families of alcoholics is controlled by a single major gene.
Alcohol and alcoholism (Oxford, Oxfordshire). Supplement (1991-01-01)
E J Devor, C R Cloninger, P L Hoffman, B Tabakoff
ABSTRACT
The stimulated activity of platelet adenylate cyclase (AC) has been shown to be significantly lower among alcoholics compared with controls. In this report we demonstrate that stimulated platelet AC activity is under the control of a single major gene locus. Thus a sensitive biological indicator of alcoholism has been shown to be highly heritable.
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%