Merck
  • Model studies on protein glycation: influence of cysteine on the reactivity of arginine and lysine residues toward glyoxal.
Model studies on protein glycation: influence of cysteine on the reactivity of arginine and lysine residues toward glyoxal.
Annals of the New York Academy of Sciences (2008-05-02)
Uwe Schwarzenbolz, Susann Mende, Thomas Henle
ABSTRACT
Mixtures of N alpha-hippurylarginin, N alpha-hippuryllysine, and glyoxal were incubated in the absence and presence of N alpha-acetylcysteine in order to assess the individual reactivity of these nucleophilic amino acid residues. The incubations were performed under atmospheric and high hydrostatic pressure (400 MPa), and, at the same time, beta-casein was reacted with glyoxal. The results showed that arginine is the main partner for glyoxal in the absence of cysteine, whereas a lysine derivatization was not apparent. In the presence of cysteine, however, arginine was almost completely protected from the reaction, whereas a noticeable formation of lysine derivatives, mainly carboxymethyllysine, was observed. Based on these findings, a reaction mechanism is proposed to explain the influence of cysteine on the reaction.
Sigma-Aldrich
Hippuryl-Lys
Sigma-Aldrich
Hippuryl-Arg, carboxypeptidase substrate