Merck
  • Urea derivatives are highly active catalysts for the base-mediated generation of terminal epoxides from aldehydes and trimethylsulfonium iodide.
Urea derivatives are highly active catalysts for the base-mediated generation of terminal epoxides from aldehydes and trimethylsulfonium iodide.
Organic & biomolecular chemistry (2008-04-04)
Sarah A Kavanagh, Alessandro Piccinini, Eimear M Fleming, Stephen J Connon
ABSTRACT
N,N'-Diarylureas have been shown to efficiently catalyse sulfonium ylide-mediated aldehyde epoxidation reactions for the first time. These processes are of broad scope and can be coupled with a subsequent Cu(II) ion-catalysed Meinwald rearrangement to give an efficient and convenient protocol for aldehyde homologation without intermediate purification.
Sigma-Aldrich
Schreiner′s Catalyst
Sigma-Aldrich
Trimethylsulfonium iodide, 98%