Merck
  • Stable, high-efficiency ionic-liquid-based mesoscopic dye-sensitized solar cells.
Stable, high-efficiency ionic-liquid-based mesoscopic dye-sensitized solar cells.
Small (Weinheim an der Bergstrasse, Germany) (2007-11-22)
Daibin Kuang, Cedric Klein, Zhipan Zhang, Seigo Ito, Jacques-E Moser, Shaik M Zakeeruddin, Michael Grätzel
ABSTRACT
Efficient and stable mesoscopic dye-sensitized solar cells (DSCs) introducing a low-viscosity binary ionic liquid (1-propyl-3-methyl-imidazolium iodide (PMII) and 1-ethyl-3-methyl-imidazolium tetracyanoborate (EMIB(CN)(4))) electrolyte in combination with a new high-molar-extinction-coefficient ruthenium complex, Ru(2,2'-bipyridine-4,4'-dicarboxylic acid)(4,4'-bis(2-(4-tert-butyloxy -phenyl)ethenyl) -2,2'-bipyridine) (NCS)(2), are demonstrated. The dependence of photovoltaic performance, charge transport and electron lifetime on the composition of the binary ionic-liquid electrolyte with different ratios of PMII/EMIB(CN)(4) were investigated by electrochemical impedance and photovoltage transient techniques. A photovoltaic conversion efficiency of 7.6 % was obtained under simulated full sunlight illumination, which is a record for solvent-free DSCs. These devices exhibit excellent stability at 80 degrees C in the dark or under visible-light soaking at 60 degrees C during 1000 h of accelerated tests.
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG