Merck
  • Advantages of synthesizing trans-1,2-cyclohexanediol in a continuous flow microreactor over a standard glass apparatus.
Advantages of synthesizing trans-1,2-cyclohexanediol in a continuous flow microreactor over a standard glass apparatus.
The Journal of organic chemistry (2007-11-16)
Andreas Hartung, Mark A Keane, Arno Kraft
ABSTRACT
Comparison is made between the preparation of trans-1,2-cyclohexanediol in standard glassware (conventional batch production) and in a microreactor (continuous flow production). The reaction sequence involved two exothermic steps where the standard procedure demands slow reagent addition and careful temperature control. In the microreactor, the reaction could be carried out safely with up to 3 times higher reagent concentration. Synthetic benefits were a faster reaction rate and a higher purity product free of colored impurities (a feature of the batch procedure).
Sigma-Aldrich
trans-1,2-Cyclohexanediol, 98%
Sigma-Aldrich
cis-1,2-Cyclohexanediol, 99%