Merck
  • Thin-layer chromatography of urinary 17-oxosteroids using dansylhydrazine as a prelabelling reagent.
Thin-layer chromatography of urinary 17-oxosteroids using dansylhydrazine as a prelabelling reagent.
Journal of chromatography (1991-10-04)
Z Tomsová
ABSTRACT
This paper describes a modification of a high-performance liquid chromatographic method for measurement of 17-oxosteroids in biological fluids for use with thin-layer chromatography and fluorometric scanning detection. After extraction from urine samples with Separon-C18 microcolumns, free oxosteroids were labelled with dansylhydrazine in acetonitrile-acetic acid and chromatographed on silica gel F-254 plates with the solvent system chloroform-methanol (97:3). Linearity of fluorescence detection (Shimadzu CS-9000 densitometer) was obtained between 30 and 1000 ng.
Sigma-Aldrich
Dansylhydrazine, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Dansylhydrazine, 98%