Merck
  • Enantioselective nickel-catalyzed cross-coupling reactions of trialkynylindium reagents with racemic secondary benzyl bromides.
Enantioselective nickel-catalyzed cross-coupling reactions of trialkynylindium reagents with racemic secondary benzyl bromides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-10-12)
Jorge Caeiro, José Pérez Sestelo, Luis A Sarandeses
ABSTRACT
The first enantioselective sp-sp3 cross-coupling reaction between alkynyl organometals and racemic benzyl bromides is reported. The coupling is performed at room temperature by using NiBr2diglyme and (S)-(iPr)-Pybox as the catalytic system and trialkynylindium reagents as nucleophiles. The reaction is stereoconvergent, both enantiomers of the racemic benzyl bromide are converted into one enantiomer of the product, and stereospecific. The reaction takes place efficiently in good yields and with high atom economy, as the triorganoindium reagents transfer the three organic groups attached to indium (only 40 mol % of R3In is used).
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%