Merck
  • Determination of pesticides in apples by liquid chromatography with electrospray ionization tandem mass spectrometry and estimation of measurement uncertainty.
Determination of pesticides in apples by liquid chromatography with electrospray ionization tandem mass spectrometry and estimation of measurement uncertainty.
Journal of AOAC International (2007-05-04)
Jian Wang, David Wotherspoon
ABSTRACT
A liquid chromatography/electrospray ionization-tandem mass spectrometry (LC/ESI-MS/MS) method was developed and validated to determine 90 pesticides in apple samples. MS/MS data acquisition was achieved by applying multiple-reaction monitoring of 2 fragment ion transitions to provide a high degree of sensitivity and selectivity for both quantitation and confirmation. Matrix-matched standard calibration curves with the use of isotopically labeled standards (or a chemical analog) as internal standards were used to achieve the best accuracy for the method. Both a conventional method validation procedure and a designed experiment were applied to study the accuracy and precision of the method. A compiled computer program that provided a semiautomated procedure for handling a large number of calculations was used to calculate the overall recovery, intermediate precision, and measurement uncertainty (MU). In general, the overall recoveries from samples spiked at levels of 10, 50, and 80 microg/kg, ranged from 60.8 to 121.1%, intermediate precision was <10%, and MUs were <30%. Poor accuracy and/or repeatability was observed for pyridate and etofenprox. The method limits of detection based on a signal-to-noise ratio of >3 were usually <1 microg/kg (ppb), except those for aldicarb sulfoxide and pyridaphenthion, which were about 5 microg/kg.
Product Description
Supelco
Etofenprox, PESTANAL®, analytical standard