Merck
  • A new analytical material-enhanced laser desorption ionization (MELDI) based approach for the determination of low-mass serum constituents using fullerene derivatives for selective enrichment.
A new analytical material-enhanced laser desorption ionization (MELDI) based approach for the determination of low-mass serum constituents using fullerene derivatives for selective enrichment.
Journal of proteome research (2007-01-06)
Rainer M Vallant, Zoltan Szabo, Lukas Trojer, Muhammad Najam-ul-Haq, Matthias Rainer, Christian W Huck, Rania Bakry, Günther K Bonn
ABSTRACT
60]fullerene derivatives (dioctadecyl methano[60]fullerene, [60]fullerenoacetic acid, and IDA-[60]fullerene) were prepared and subjected to a comprehensive characterization study including protein binding properties and capacity. These fullerene derivatives were successfully applied as material-enhanced laser desorption/ionization (MELDI) carrier materials. It is shown that diverse functionalities result in characteristic human serum peak patterns (m/z 2000-20 000) in terms of signal intensity as well as the number of detectable masses. In addition, the fullerene derivatives clearly provided differences in the low molecular weight mass region (m/z 1000-4000) after elution of the adsorbed serum constituents, and [60]fullerenoacetic acid was the most effective carrier material. Novel high-speed, monolithic, high-resolution capillary columns, prepared by thermally initiated copolymerization of methylstyrene (MSt) and 1,2-bis(p-vinylphenyl)ethane (BVPE) were employed for eluate separation and target spotting. Thus, serum compounds in the low-mass range were successfully fractionated and subjected to MALDI-MS/MS analysis. This contribution, hence, proposes a new "top-down" strategy for proteome research enabling protein profiling as well as biomarker identification in the low-mass range using selective enrichment, high-resolution separation, and offline MALDI-MS/MS evaluation.
Sigma-Aldrich
Methylstyrene, 60% meta, 40% para and 1% ortho, 99%, contains ~50 ppm 4-tert-butylcatechol as inhibitor