Merck
  • A fluorescent electrophilic reagent, 9-fluorenone-4-carbonyl chloride (FCC), for the enantioresolution of amino acids on a teicoplanin phase under the elution of the methanol-based solvent mixture.
A fluorescent electrophilic reagent, 9-fluorenone-4-carbonyl chloride (FCC), for the enantioresolution of amino acids on a teicoplanin phase under the elution of the methanol-based solvent mixture.
Amino acids (2006-10-28)
T-J Hsien, S Chen
ABSTRACT
A fluorescent electrophilic reagent, 9-fluorenone-4-carbonyl chloride (FCC), is chosen to functionalize amino acids in alkaline medium before their HPLC resolution. FCC reacts with both primary and secondary amino acids to produce stable and highly fluorescent derivatives suitable for sensitive and efficient chromatographic determination and resolution on a teicoplanin chiral stationary phase (CSP) using the methanol-based solvent mixture as the mobile phase. The detection limit is in the picomole range and approximately 0.01% of the D: -enantiomer in an excess of the L: -enantiomer is detectable. However, the resolution is not reproducible under the elution of either the water- or the acetonitrile-based mobile phase. The increase in solubility of analyte in the mobile phase seems to be responsible. Upon comparison under the optimal chromatographic conditions, the resolution is better than that for the 9-fluorenylmethyl chloroformate (FMOC) or 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate (AQC) derivatives reported previously.
Sigma-Aldrich
9-Fluorenone, 98%