Merck
  • Asymmetric synthesis of 4,5,6- and 3,4,5,6-substituted azepanes by a highly diastereoselective and enantioselective lithiation-conjugate addition sequence.
Asymmetric synthesis of 4,5,6- and 3,4,5,6-substituted azepanes by a highly diastereoselective and enantioselective lithiation-conjugate addition sequence.
Journal of the American Chemical Society (2006-02-16)
Suk Joong Lee, Peter Beak
ABSTRACT
Asymmetric syntheses of 4,5,6- and 3,4,5,6-substituted azepanes have been achieved by highly diastereoselective and enantioselective (-)-sparteine-mediated asymmetric lithiation-conjugate additions of N-Boc-N-(p-methoxyphenyl)-2,3-substituted allylamines to a beta-aryl alpha,beta-unsaturated ester followed by hydrolysis, cyclization, and reduction. Access to the enantiomeric adduct is provided by an invertive lithiation-stannylation-lithiation sequence.
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mL
In Stock
Details...
US$41.60
100 mL
In Stock
Details...
US$49.10
500 mL
In Stock
Details...
US$88.40