Merck
  • Preconcentration and determination of mercury(II) at a chemically modified electrode containing 3-(2-thioimidazolyl)propyl silica gel.
Preconcentration and determination of mercury(II) at a chemically modified electrode containing 3-(2-thioimidazolyl)propyl silica gel.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-12-02)
Newton L Dias Filho, Devaney R do Carmo, Laércio Caetano, André H Rosa
ABSTRACT
A mercury-sensitive chemically modified graphite paste electrode was constructed by incorporating modified silica gel into a conventional graphite paste electrode. The functional group attached to the (3-chloropropyl) silica gel surface was 2-mercaptoimidazole, giving a new product denoted by 3-(2-thioimidazolyl)propyl silica gel, which is able to complex mercury ions. Mercury was chemically adsorbed on the modified graphite paste electrode containing 3-(2-thioimidazolyl)propyl silica (TIPSG GPE) by immersion in a Hg(II) solution, and the resultant surface was characterized by cyclic and differential pulse anodic stripping voltammetry. One cathodic peak at 0.1 V and other anodic peak at 0.34 V were observed on scanning the potential from -0.1 to 0.8 V (0.01 M KNO3; v = 2.0 mV s(-1) vs. Ag/AgCl). The anodic peak at 0.34 V show an excellent sensitivity for Hg(II) ions in the presence of several foreign ions. A calibration graph covering the concentration range from 0.02 to 2 mg L(-1) was obtained. The detection limit was estimated to be 5 microg L(-1). The precision for six determinations of 0.05 and 0.26 mg L(-1) Hg(II) was 3.0 and 2.5% (relative standard deviation), respectively. The method can be used to determine the concentration of mercury(II) in natural waters contaminated by this metal.
Sigma-Aldrich
2-Mercaptoimidazole, 98%