Merck
  • Bioavailability of 2,3',4,4',5-pentachlorobiphenyl (PCB118) and 2,2',5,5'-tetrachlorobiphenyl (PCB52) from soils using a rat model and a physiologically based extraction test.
Bioavailability of 2,3',4,4',5-pentachlorobiphenyl (PCB118) and 2,2',5,5'-tetrachlorobiphenyl (PCB52) from soils using a rat model and a physiologically based extraction test.
Toxicology (2005-09-21)
Xinzhu Pu, Linda S Lee, Raymond E Galinsky, Gary P Carlson
ABSTRACT
The bioavailability of coplanar 2,3',4,4',5-pentachlorobiphenyl (PCB118) and nonplanar 2,2',5,5'-tetrachlorobiphenyl (PCB52) from soils representing a range in organic carbon (OC), clay content and pH were investigated using an in vivo rat model and an in vitro physiologically based extraction test (PBET) to assess the role of soil and chemical properties on bioavailabilty. Affinity to soil and persistence of PCBs have been shown to increase with increasing soil organic carbon (OC) content, PCB chlorination, and PCB coplanarity. In the in vivo tests for both PCB118 and PCB52, the AUCs following iv injection were significantly higher than the AUCs for all soil groups, indicating that the soil matrix can reduce the absolute bioavailability of PCB118 and PCB52. However, no significant differences were detected between soils of different properties. In the in vitro PBET, significant differences in the mobilization of PCB118 and PCB52 were observed among soils, and PCBs had the least mobilization from the soil with the highest OC content consistent with hydrophobic partitioning theory. Also, significantly less PCB118 was mobilized relative to PCB52 in the PBET assay, showing the potential impact of spatial orientation and chlorine content on bioavailability. No correlation between the in vitro PBET and the in vivo rat model was observed for the PCBs. Although the in vitro PBET and related assays may serve as an indicator of bioavailability, it is likely to underestimate what can be released from a soil in an in vivo assay.
Product Description
2,2′,5,5′-Tetrachlorobiphenyl (IUPAC No. 52), BCR®, certified reference material
Pricing and availability is not currently available.
Supelco
PCB No 52, analytical standard
Pricing and availability is not currently available.