Merck
  • Preparation and characterization of carbonated barium-calcium hydroxyapatite solid solutions.
Preparation and characterization of carbonated barium-calcium hydroxyapatite solid solutions.
Journal of colloid and interface science (2005-06-02)
Akemi Yasukawa, Eiichi Ueda, Kazuhiko Kandori, Tatsuo Ishikawa
ABSTRACT
Particles of carbonated barium-calcium hydroxyapatite solid solutions (BaCaHap) with different Ba/(Ba+Ca) (X(Ba)) atomic ratios were prepared by a wet method at 100 degrees C and characterized by various means. The crystal phases and structures of the products strongly depended on the composition of the starting solution, that is, the Ba/(Ba+Ca) atomic ratio ([X(Ba)]) and H3PO4 concentration ([H3PO4]) in the solution. BaCaHap with X(Ba)0.43 could be prepared at [X(Ba)]0.7 by changing [H3PO4], but could never be obtained at [X(Ba)]=0.8-0.95 regardless of [H3PO4]. The carbonated calcium hydroxyapatite particles prepared at [X(Ba)]=0 were fine and short rod-shaped particles (ca. 14x84 nm). With increasing [X(Ba)] from 0 to 0.8, the particles obtained became large spherical agglomerates. The carbonated barium hydroxyapatite particles formed at [X(Ba)]=1 were long rod-shaped agglomerates (ca. 0.2x2 microm) of fine primary particles. The amount of CO2 adsorbed irreversibly on a series of BaCaHaps showed a minimum at (Ba+Ca)/(P+C) atomic ratio of around 1.56, which agreed well with the minimum cation/P ratio obtained for the other hydroxyapatites, as already reported.
Sigma-Aldrich
Barium carbonate, 99.98% trace metals basis
Sigma-Aldrich
Barium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Barium carbonate, ACS reagent, ≥99%