Merck
  • Effects of ethanol and its fluorinated analogues on the calcium ATPase of sarcoplasmic reticulum.
Effects of ethanol and its fluorinated analogues on the calcium ATPase of sarcoplasmic reticulum.
Biochemistry international (1992-05-01)
F C Bennett, S D Gates, G F King, B C Shanley, R Smith
ABSTRACT
The effects of ethanol, monofluoro- and trifluoroethanol on the hydrolysis of ATP and coupled Ca2+ translocation by sarcoplasmic reticulum Ca(2+)-ATPase (EC 3.6.1.38) were measured. All three alcohols had parallel effects on the enzyme activities at concentrations up to 200 mM, suggesting a similar mechanism of action. 19F nuclear magnetic resonance spectroscopy was used to investigate their site of action in the purified enzyme.