Merck
  • Study on the retention behaviour of metal-tetraphenyl porphine chelates in reversed-phase high performance liquid chromatography.
Study on the retention behaviour of metal-tetraphenyl porphine chelates in reversed-phase high performance liquid chromatography.
Se pu = Chinese journal of chromatography (2003-01-25)
Z H Shi, Y H Wang, C G Fu
ABSTRACT
The retention behaviour of metal chelates of alpha,beta,gamma,delta-tetraphenylporphine (TPP) was studied in reversed-phase high performance liquid chromatography (RP-HPLC). On C18 column, with various organic solvents as mobile phases, the retention of the chelates tends to increase in the following order: ZnTPP < TPP < NiTPP < CuTPP. This retention behaviour can get a good explanation from Horvath's "solvophobic theory". With the assumption that the electrostatic field strength of metal chelates can be measured by the ratio of electronegativity vs. ionic radius (EN/ri) of central metal ion, EN/ri was established to be an index for the retention behaviour of metal-TPP chelates. It was found that lnk' is in good linear relationship with the EN/ri value.
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, ≥99%
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine, 97%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
5,10,15,20-Tetraphenyl-21H,23H-porphine zinc