Merck
  • Ion mobility spectrometry of hydrazine, monomethylhydrazine, and ammonia in air with 5-nonanone reagent gas.
Ion mobility spectrometry of hydrazine, monomethylhydrazine, and ammonia in air with 5-nonanone reagent gas.
Analytical chemistry (1993-01-01)
G A Eiceman, M R Salazar, M R Rodriguez, T F Limero, S W Beck, J H Cross, R Young, J T James
ABSTRACT
Hydrazine (HZ) and monomethylhydrazine (MMH) in air were monitored continuously using a hand-held ion mobility spectrometer equipped with membrane inlet, 63Ni ion source, acetone reagent gas, and ambient temperature drift tube. Response characteristics included detection limit, 6 ppb; linear range, 10-600 ppb; saturated response, >2 ppm; and stable response after 15-30 min. Ammonia interfered in hydrazines detection through a product ion with the same drift time as that for MMH and HZ. Acetone reagent gas was replaced with 5-nonanone to alter drift times of product ions and separate ammonia from MMH and HZ. Patterns in mobility spectra, ion identifications from mass spectra, and fragmentation cross-sections from collisional-induced dissociations suggest that drift times are governed by ion-cluster equilibria in the drift region of the mobility spectrometer. Practical aspects including calibration, stability, and reproducibility are reported from the use of a hand-held mobility spectrometer on the space shuttle Atlantis during mission STS-37.
Sigma-Aldrich
5-Nonanone, 98%