Merck
  • New preconditioning strategy for the determination of inorganic anions with capillary zone electrophoresis using indirect UV detection.
New preconditioning strategy for the determination of inorganic anions with capillary zone electrophoresis using indirect UV detection.
Journal of chromatography. A (2000-09-29)
G Raber, H Greschonig
ABSTRACT
It is widely accepted that preconditioning procedures are indispensable in capillary electrophoresis in order to achieve reproducibility of migration times and peak areas. Several preconditioning strategies have been employed for electrophoretic determinations of inorganic anions using indirect UV detection including simple flushing with buffer or alkaline or acid pre-rinsing followed by flushing with electrolyte. We investigated the influence of various preconditioning strategies on the reproducibility of migration times and peak areas of inorganic anions. The electrolyte systems for indirect UV detection were based on pyromellitic acid and chromic acid respectively as UV absorbing probes and hexamethonium hydroxide as electroosmatic flow modifier. Preconditioning agents under investigation were electrolyte buffer, NaOH, HCl and the free acids of the UV absorbing probes. Investigations showed that reproducibility of migration times and peak areas can be significantly improved by acid pre-rinsing using the corresponding acid of the UV absorbing probes compared to preconditioning by flushing the capillary with buffer. In contrast to acid pre-rinsing using hydrochloric acid no interfering signals within the migration time window of inorganic anions under investigation can be observed. The optimized preconditioning procedure yields relative standard deviations of migration times less than 0.25% (n = 10). Relative standard deviations of corrected peak areas were below 5% applying acid preconditioning using pyromellitic acid.
Sigma-Aldrich
1,2,4,5-Benzenetetracarboxylic acid, 96%