Merck
  • Solid-phase synthesis of benzisothiazolones as serine protease inhibitors.
Solid-phase synthesis of benzisothiazolones as serine protease inhibitors.
Bioorganic & medicinal chemistry letters (1999-04-14)
K L Yu, R Civiello, D G Roberts, S M Seiler, N A Meanwell
ABSTRACT
An efficient solid-phase synthesis of benzisothiazolone-1,1-dioxide-based serine protease inhibitors involving alkylation of carboxylic acids with N-(bromomethyl)benzisothiazolone-1,1-dioxide has been developed. An example using this procedure for preparation of a library of human mast cell tryptase inhibitors is described.
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
Supelco
1,2-Benzisothiazol-3(2H)-one, analytical standard