Merck
Isolation and characterization of the human hepatic lipase gene.
The Journal of biological chemistry (1990-04-25)
D Ameis, G Stahnke, J Kobayashi, J McLean, G Lee, M Büscher, M C Schotz, H Will
ABSTRACT
Overlapping bacterial phage and cosmid genomic clones were isolated spanning an area of approximately 60 kilobases that contains the human hepatic lipase (HL) gene. It is composed of 9 exons spanning approximately 35 kilobases of DNA. The entire coding regions, the 5'-flanking sequences, and the exon-intron junctions were sequenced. The intron positions correspond to those of human lipoprotein lipase and canine pancreatic lipase, supporting the concept that these genes constitute a dispersed gene family of lipases and have evolved by duplication of a common ancestral gene. A region of the HL gene, which displays a significant homology with various other lipolytic enzymes and contains the putative catalytic site serine residue of HL, was encoded by exon 4. A major transcription start site of the human HL gene was located by primer extension analysis, 43 nucleotides upstream of the translation initiation codon. Two possible promoter elements were located 25 and 63 nucleotides upstream of the transcription initiation site: a "TATA" box-like sequence, TAATA, and a sequence found in the promoter region of many liver-specific genes, AGGTTAATTATTAAT. In addition, sequences homologous to glucocorticoid and cAMP-responsive elements were identified in the 5'-nontranscribed region.
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg