Merck
  • Adeno-associated viral (AAV) serotype 5 vector mediated gene delivery of endothelin-converting enzyme reduces Abeta deposits in APP + PS1 transgenic mice.
Adeno-associated viral (AAV) serotype 5 vector mediated gene delivery of endothelin-converting enzyme reduces Abeta deposits in APP + PS1 transgenic mice.
Molecular therapy : the journal of the American Society of Gene Therapy (2008-07-31)
Niki C Carty, Kevin Nash, Daniel Lee, Mary Mercer, Paul E Gottschall, Craig Meyers, Nicholas Muzyczka, Marcia N Gordon, Dave Morgan
ABSTRACT
Reduction of Abeta deposition is a major therapeutic strategy in Alzheimer's disease (AD). The concentration of Abeta in the brain is modulated not only by Abeta production but also by its degradation. One of the proteases involved in the degradation of Abeta peptides is endothelin-converting enzyme (ECE). In this study, we investigated the effects of an intracranial administration of a seroptype 5 recombinant adeno-associated viral vector (rAAV) containing the ECE-1 synthetic gene on amyloid deposition in amyloid precursor protein (APP) plus presenilin-1 (PS1) transgenic mice. The rAAV vector was injected unilaterally into the right anterior cortex and hippocampus of 6-month-old mice, while control mice received an AAV vector expressing green fluorescent protein (GFP). Immunohistochemical testing for the hemagglutinin (HA) tag appended to ECE revealed strong expression in areas surrounding the injection sites but minimal expression in the contralateral regions. Immunohistochemical tests showed that Abeta decreases in the anterior cortex and hippocampus in mice receiving the ECE synthetic gene. Further, decreases in Congo red positive deposits were also observed in both regions. These results indicate that increasing the expression of beta-amyloid degrading enzymes through gene therapy is a promising approach to the treatment of AD.
Sigma-Aldrich
SM-19712 hydrate, ≥98% (HPLC)