Merck
[The β-amylase polymorphism of winter common wheat grains].
Genetika (2012-05-10)
V P Netsvetaev, O V Akinshina, L S Bondarenko, I P Motorina
ABSTRACT
The polymorphism of winter common wheat with respect to β-amylase isoenzymes has been analyzed using electrophoresis in polyacrylamide gel (PAAG) buffered with a Tris-glycine system (pH 8.3). Seven β-amylase isoenzymes have been found in wheat cultivars and the breeding stock. Isoenzymes A, B, and C are the most frequent in Russian and Ukrainian cultivars (51.7 4.7, 30.7 3.8, and 11.9 2.5%, respectively). Two alleles of the β-Amy-D1 locus of the long arm of chromosome 4D have been identified. The substrate-enzyme affine effect can be used to locate the zones of activity of this enzyme by means of staining for proteins. It has been determined that β-amylase zymotypes may play a role in the aggregating capacity of the grain protein complex via the formation of S-S bonds.
Sigma-Aldrich
β-Amylase from barley, Type II-B, 20-80 units/mg protein (biuret)
Sigma-Aldrich
β-Amylase from sweet potato, Type I-B, ammonium sulfate suspension, ≥750 units/mg protein (E1%/280)
Sigma-Aldrich
β-Amylase from sweet potato