Merck
  • Influence of dosing regimen on alprazolam and metabolite serum concentrations and tolerance to sedative and psychomotor effects.
Influence of dosing regimen on alprazolam and metabolite serum concentrations and tolerance to sedative and psychomotor effects.
Psychopharmacology (1987-01-01)
R B Smith, P D Kroboth
ABSTRACT
The relationships between alprazolam and metabolite concentrations and CNS effects were determined in a double-blind placebo controlled four-way crossover trial in 16 normal male volunteers. Active drug treatments consisted of 4-day regimens of 4 mg alprazolam PO daily as 2 mg bid., 1 mg qid, and 0.25 mg each hour. On days 1 and 4, the kinetics, sedative and psychomotor effects were evaluated. Plasma concentrations of the 4- and alpha-hydroxy metabolites of alprazolam were less than 10% of unchanged alprazolam levels on both days. Accumulation of these metabolites and alprazolam was dependent on alprazolam half-life (11.6 h). Acute and chronic tolerance to the sedative and psychomotor effects was observed with all active drug treatments. All alprazolam treatments resulted in significantly greater sedation than placebo on days 1 and 4. However, on day 4, sedation was 16-36% less than observed on day 1, despite plasma concentrations 1.4-2.76 times the day 1 concentrations. Sedation from alprazolam was reduced in each successive study phase, suggesting a tolerance which was sustained during the 10-day washout between phases. By day 4, psychomotor performance was not different from placebo, indicating more complete development of tolerance than occurred for the sedative effect. Sedation and psychomotor impairment on day 1 were greatest with 2 mg alprazolam bid. During the initiation of therapy, the patient will likely experience less sedation and psychomotor impairment with smaller, more frequent doses. Since tolerance develops to these effects, the advantage of more frequent dosing regimen dissipates by the 4th day.
Sigma-Aldrich
α-Hydroxyalprazolam, powder, ≥98% (TLC)