Merck
  • Initial evaluation of the antitumour activity of KGP94, a functionalized benzophenone thiosemicarbazone inhibitor of cathepsin L.
Initial evaluation of the antitumour activity of KGP94, a functionalized benzophenone thiosemicarbazone inhibitor of cathepsin L.
European journal of medicinal chemistry (2012-11-22)
Gustavo E Chavarria, Michael R Horsman, Wara M Arispe, G D Kishore Kumar, Shen-En Chen, Tracy E Strecker, Erica N Parker, David J Chaplin, Kevin G Pinney, Mary Lynn Trawick
ABSTRACT
Kinetic analysis of the mode of inhibition of cathepsin L by KGP94, a lead compound from a privileged library of functionalized benzophenone thiosemicarbazone derivatives, demonstrated that it is a time-dependent, reversible, and competitive inhibitor of the enzyme. These results are consistent with the formation of a transient covalent bond, and are supported by molecular modeling that places the thiocarbonyl of the inhibitor in proximity to the thiolate moiety of the enzyme active site Cys25. KGP94 significantly decreased the activity of cathepsin L toward human type I collagen, and impeded both migration and invasion of MDA-MB-231 human breast cancer cells. Growth retardation was achieved in vivo against both recently implanted and established tumours using a C3H mouse mammary carcinoma model.
Sigma-Aldrich
Cathepsin L from human liver, ≥0.5 units/mg protein, solution