Merck
  • Genetic mapping of an autosomal recessive postaxial polydactyly type A to chromosome 13q13.3-q21.2 and screening of the candidate genes.
Genetic mapping of an autosomal recessive postaxial polydactyly type A to chromosome 13q13.3-q21.2 and screening of the candidate genes.
Human genetics (2011-08-31)
Umm-e-Kalsoom, Sulman Basit, Syed Kamran-ul-Hassan Naqvi, Muhammad Ansar, Wasim Ahmad
ABSTRACT
Postaxial Polydactyly (PAP) is characterized by fifth digit duplication in hands and/or feet. Two types of PAP including PAP-A, representing the development of well-formed extra digit, and PAP-B, representing the presence of rudimentary fifth digit, have been described. Both isolated and syndromic forms of PAP have been reported. Isolated forms of PAP usually segregate as an autosomal dominant trait and to date four loci have been identified. In the present study, we have described mapping of the first locus of autosomal recessive PAP type A on chromosome 13q13.3-13q21.2 in a consanguineous Pakistani family. Using polymorphic microsatellite markers, the disease locus was mapped to a 17.87-cM (21.13 Mb) region flanked by markers D13S1288 and D13S632, on chromosome 13q13.3-13q21.2. A maximum multipoint LOD score of 3.84 was obtained with several markers along the disease interval. DNA sequence analysis of exons and splice-junction sites of ten candidate genes (CHM-I, TSC22D1, FOXO1, DIAPH3, CCDC122, CKAP2, SUGT1, RANKL, LPAR6, C13ORF31) did not reveal potentially causal variants.
Sigma-Aldrich
GenElute Blood Genomic DNA Kit, sufficient for 350 purifications
Sigma-Aldrich
GenElute Blood Genomic DNA Kit, sufficient for 70 purifications