Merck
Loss of Calponin 2 causes premature ovarian insufficiency in mice.
Journal of ovarian research (2024-02-10)
Tzu-Bou Hsieh, Jian-Ping Jin
ABSTRACT
Premature ovarian insufficiency (POI) is a condition defined as women developing menopause before 40 years old. These patients display low ovarian reserve at young age and difficulties to conceive even with assisted reproductive technology. The pathogenesis of ovarian insufficiency is not fully understood. Genetic factors may underlie most of the cases. Actin cytoskeleton plays a pivotal role in ovarian folliculogenesis. Calponin 2 encoded by the Cnn2 gene is an actin associated protein that regulates motility and mechanical signaling related cellular functions. The present study compared breeding of age-matched calponin 2 knockout (Cnn2-KO) and wild type (WT) mice and found that Cnn2-KO mothers had significantly smaller litter sizes. Ovaries from 4 weeks old Cnn2-KO mice showed significantly lower numbers of total ovarian follicles than WT control with the presence of multi-oocyte follicles. Cnn2-KO mice also showed age-progressive earlier depletion of ovarian follicles. Cnn2 expression is detected in the cumulus cells of the ovarian follicles of WT mice and colocalizes with actin stress fiber, tropomyosin and myosin II in primary cultures of cumulus cells. The findings demonstrate that the loss of calponin 2 impairs ovarian folliculogenesis with premature depletion of ovarian follicles. The role of calponin 2 in ovarian granulosa cells suggests a molecular target for further investigations on the pathogenesis of POI and for therapeutic development.
Sigma-Aldrich
Monoclonal Anti-Rabbit Immunoglobulins−Alkaline Phosphatase antibody produced in mouse, clone RG-16, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DDX4 (Vasa) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution