Merck
Exposure to low-dose cadmium induces testicular ferroptosis.
Ecotoxicology and environmental safety (2022-03-11)
Lijuan Xiong, Bin Zhou, Jamie L Young, Kupper Wintergerst, Lu Cai
ABSTRACT
As an environmental pollutant, cadmium (Cd) has been widely reported to induce male infertility due to its gonadotoxicity. However, the specific mechanism of Cd-induced testicular damage remains unclear. We investigated whether Cd causes testicular injury through ferroptosis. Male C57BL/6 J mice were exposed to 0, 0.5, or 5 ppm Cd via drinking water, starting in utero, and continuing through 24 weeks post-weaning. The results showed that Cd accumulated in the testes in a dose-dependent manner. Cd exposure at a concentration of 5 ppm, but not 0.5 ppm, caused a mass loss and detachment of germ cells, as well as a decreased meiotic index and testis weight. Exposure to 5 ppm Cd caused iron accumulation, increased levels of malondialdehyde (MDA) and nitro tyrosine (3-NT), and decreased expression of Nrf2, HO-1 and SOD2. We also found that exposure to 5 ppm Cd significantly decreased the expression of SLC7A11, a marker of ferroptosis in mice, along with the expression of SLC40A1 mRNA and ferritin heavy chain (FTH) protein, whereas there was no obvious change in the mRNA expression of Tfrc, ZIP8, ZIP14, and NCOA4. These findings indicate that 5 ppm Cd exposure increased testicular ferroptosis, which may be attributed to the reduction of stored iron export.
Sigma-Aldrich
Anti-Nitrotyrosine antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution