Merck
  • Galectin-1 induces a tumor-associated macrophage phenotype and upregulates indoleamine 2,3-dioxygenase-1.
Galectin-1 induces a tumor-associated macrophage phenotype and upregulates indoleamine 2,3-dioxygenase-1.
iScience (2023-08-03)
Asha M Rudjord-Levann, Zilu Ye, Lise Hafkenscheid, Sabrina Horn, Renske Wiegertjes, Mathias A I Nielsen, Ming Song, Caroline B K Mathiesen, Jesse Stoop, Sean Stowell, Per Thor Straten, Hakon Leffler, Sergey Y Vakhrushev, Sally Dabelsteen, Jesper V Olsen, Hans H Wandall
ABSTRACT
Galectins are a group of carbohydrate-binding proteins with a presumed immunomodulatory role and an elusive function on antigen-presenting cells. Here we analyzed the expression of galectin-1 and found upregulation of galectin-1 in the extracellular matrix across multiple tumors. Performing an in-depth and dynamic proteomic and phosphoproteomic analysis of human macrophages stimulated with galectin-1, we show that galectin-1 induces a tumor-associated macrophage phenotype with increased expression of key immune checkpoint protein programmed cell death 1 ligand 1 (PD-L1/CD274) and immunomodulator indoleamine 2,3-dioxygenase-1 (IDO1). Galectin-1 induced IDO1 and its active metabolite kynurenine in a dose-dependent manner through JAK/STAT signaling. In a 3D organotypic tissue model system equipped with genetically engineered tumorigenic epithelial cells, we analyzed the cellular source of galectin-1 in the extracellular matrix and found that galectin-1 is derived from epithelial and stromal cells. Our results highlight the potential of targeting galectin-1 in immunotherapeutic treatment of human cancers.
Sigma-Aldrich
Anti-IDO Antibody, clone 1F8.2, clone 1F8.2, from mouse