Merck
  • Lingual Denervation Improves the Efficacy of Anti-PD-1 Immunotherapy in Oral Squamous Cell Carcinomas by Downregulating TGFβ Signaling.
Lingual Denervation Improves the Efficacy of Anti-PD-1 Immunotherapy in Oral Squamous Cell Carcinomas by Downregulating TGFβ Signaling.
Cancer research communications (2024-02-07)
Zhuo-Ying Tao, Leilei Wang, Wang-Yong Zhu, Gao Zhang, Yu-Xiong Su
ABSTRACT
Intratumoral nerve infiltration relates to tumor progression and poor survival in oral squamous cell carcinoma (OSCC). How neural involvement regulates antitumor immunity has not been well characterized. This study aims to investigate molecular mechanisms of regulating tumor aggressiveness and impairing antitumor immunity by nerve-derived factors. We performed the surgical lingual denervation in an immunocompetent mouse OSCC model to investigate its effect on tumor growth and the efficacy of anti-PD-1 immunotherapy. A trigeminal ganglion neuron and OSCC cell coculture system was established to investigate the proliferation, migration, and invasion of tumor cells and the PD-L1 expression. Both the neuron-tumor cell coculture in vitro model and the OSCC animal model were explored. Lingual denervation slowed down tumor growth and improved the efficacy of anti-PD-1 treatment in the OSCC model. Coculturing with neurons not only enhanced the proliferation, migration, and invasion but also upregulated TGFβ-SMAD2 signaling and PD-L1 expression of tumor cells. Treatment with the TGFβ signaling inhibitor galunisertib reversed nerve-derived tumor aggressiveness and downregulated PD-L1 on tumor cells. Similarly, lingual denervation in vivo decreased TGFβ and PD-L1 expression and increased CD8+ T-cell infiltration and the expression of IFNγ and TNFα within tumor. Neural involvement enhanced tumor aggressiveness through upregulating TGFβ signaling and PD-L1 expression in OSCC, while denervation of OSCC inhibited tumor growth, downregulated TGFβ signaling, enhanced activities of CD8+ T cells, and improved the efficacy of anti-PD-1 immunotherapy. This study will encourage further research focusing on denervation as a potential adjuvant therapeutic approach in OSCC. This study revealed the specific mechanisms for nerve-derived cancer progression and impaired antitumor immunity in OSCC, providing a novel insight into the cancer-neuron-immune network as well as pointing the way for new strategies targeting nerve-cancer cross-talk as a potential adjuvant therapeutic approach for OSCC.
Sigma-Aldrich
Anti-TNFA antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture