Merck
  • The Force-Dependent Mechanism of an Integrin α4β7-MAdCAM-1 Interaction.
The Force-Dependent Mechanism of an Integrin α4β7-MAdCAM-1 Interaction.
International journal of molecular sciences (2023-11-25)
Youmin Su, Zhiqing Luo, Dongshan Sun, Bishan Yang, Quhuan Li
ABSTRACT
The interaction between integrin α4β7 and mucosal vascular addressin cell-adhesion molecule-1 (MAdCAM-1) facilitates the adhesion of circulating lymphocytes to the surface of high endothelial venules in inflammatory bowel diseases (IBDs). Lymphocyte adhesion is a multistep cascade involving the tethering, rolling, stable adhesion, crawling, and migration of cells, with integrin α4β7 being involved in rolling and stable adhesions. Targeting the integrin α4β7-MAdCAM-1 interaction may help decrease inflammation in IBDs. This interaction is regulated by force; however, the underlying mechanism remains unknown. Here, we investigate this mechanism using a parallel plate flow chamber and atomic force microscopy. The results reveal an initial increase in the lifetime of the integrin α4β7-MAdCAM-1 interaction followed by a decrease with an increasing force. This was manifested in a two-state curve regulated via a catch-bond-slip-bond conversion regardless of Ca2+ and/or Mg2+ availability. In contrast, the mean rolling velocity of cells initially decreased and then increased with the increasing force, indicating the flow-enhanced adhesion. Longer tether lifetimes of single bonds and lower rolling velocities mediated by multiple bonds were observed in the presence of Mg2+ rather than Ca2+. Similar results were obtained when examining the adhesion to substrates co-coated with chemokine CC motif ligand 25 and MAdCAM-1, as opposed to substrates coated with MAdCAM-1 alone. In conclusion, the integrin α4β7-MAdCAM-1 interaction occurs via ion- and cytokine-dependent flow-enhanced adhesion processes and is regulated via a catch-bond mechanism.
Sigma-Aldrich
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
Sigma-Aldrich
Anti-6-His antibody produced in rabbit, affinity isolated antibody