Merck
  • Cyclin D1-Cdk4 regulates neuronal activity through phosphorylation of GABAA receptors.
Cyclin D1-Cdk4 regulates neuronal activity through phosphorylation of GABAA receptors.
Cellular and molecular life sciences : CMLS (2023-09-09)
Neus Pedraza, Ma Ventura Monserrat, Francisco Ferrezuelo, Jordi Torres-Rosell, Neus Colomina, Federico Miguez-Cabello, Javier Picañol Párraga, David Soto, Esperanza López-Merino, Celia García-Vilela, José A Esteban, Joaquim Egea, Eloi Garí
ABSTRACT
Nuclear Cyclin D1 (Ccnd1) is a main regulator of cell cycle progression and cell proliferation. Interestingly, Ccnd1 moves to the cytoplasm at the onset of differentiation in neuronal precursors. However, cytoplasmic functions and targets of Ccnd1 in post-mitotic neurons are unknown. Here we identify the α4 subunit of gamma-aminobutyric acid (GABA) type A receptors (GABAARs) as an interactor and target of Ccnd1-Cdk4. Ccnd1 binds to an intracellular loop in α4 and, together with Cdk4, phosphorylates the α4 subunit at threonine 423 and serine 431. These modifications upregulate α4 surface levels, increasing the response of α4-containing GABAARs, measured in whole-cell patch-clamp recordings. In agreement with this role of Ccnd1-Cdk4 in neuronal signalling, inhibition of Cdk4 or expression of the non-phosphorylatable α4 decreases synaptic and extra-synaptic currents in the hippocampus of newborn rats. Moreover, according to α4 functions in synaptic pruning, CCND1 knockout mice display an altered pattern of dendritic spines that is rescued by the phosphomimetic α4. Overall, our findings molecularly link Ccnd1-Cdk4 to GABAARs activity in the central nervous system and highlight a novel role for this G1 cyclin in neuronal signalling.
Sigma-Aldrich
Anti-GABA A Receptor α4 Antibody, CT, Chemicon®, from rabbit
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution