Merck
  • Engineering ligand-specific biosensors for aromatic amino acids and neurochemicals.
Engineering ligand-specific biosensors for aromatic amino acids and neurochemicals.
Cell systems (2021-11-13)
Austin G Rottinghaus, Chenggang Xi, Matthew B Amrofell, Hyojeong Yi, Tae Seok Moon
ABSTRACT
Microbial biosensors have diverse applications in metabolic engineering and medicine. Specific and accurate quantification of chemical concentrations allows for adaptive regulation of enzymatic pathways and temporally precise expression of diagnostic reporters. Although biosensors should differentiate structurally similar ligands with distinct biological functions, such specific sensors are rarely found in nature and challenging to create. Using E. coli Nissle 1917, a generally regarded as safe microbe, we characterized two biosensor systems that promiscuously recognize aromatic amino acids or neurochemicals. To improve the sensors' selectivity and sensitivity, we applied rational protein engineering by identifying and mutagenizing amino acid residues and successfully demonstrated the ligand-specific biosensors for phenylalanine, tyrosine, phenylethylamine, and tyramine. Additionally, our approach revealed insights into the uncharacterized structure of the FeaR regulator, including critical residues in ligand binding. These results lay the groundwork for developing kinetically adaptive microbes for diverse applications. A record of this paper's transparent peer review process is included in the supplemental information.
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
Dopamine hydrochloride