Merck
  • A possible function of Nik-related kinase in the labyrinth layer of delayed delivery mouse placentas.
A possible function of Nik-related kinase in the labyrinth layer of delayed delivery mouse placentas.
The Journal of reproduction and development (2022-12-26)
Hiroshi Yomogita, Hikaru Ito, Kento Hashimoto, Akihiko Kudo, Toshiaki Fukushima, Tsutomu Endo, Yoshikazu Hirate, Yoshihiro Akimoto, Masayuki Komada, Yoshiakira Kanai, Naoyuki Miyasaka, Masami Kanai-Azuma
ABSTRACT
In mice and humans, Nik-related protein kinase (Nrk) is an X-linked gene that encodes a serine/threonine kinase belonging to GCK group 4. Nrk knockout (Nrk KO) mice exhibit delayed delivery, possibly due to defective communication between the Nrk KO conceptus and its mother. However, the mechanism of delayed labor remains largely unknown. Here, we found that in pregnant mothers with the Nrk KO conceptus, the serum progesterone (P4) and placental lactogen (PL-2) concentrations in late pregnancy were higher than those in the wild type. Moreover, we demonstrated that Nrk is expressed in trophoblast giant cells (TGCs) and syncytiotrophoblast-2 (SynT-2) in the labyrinth layer of the mouse placenta. In the human placenta, NRK is also expressed in Syn-T in villi. Both human Syn-T and mouse TGCs of the labyrinth layer are present within fetal tissues that are in direct contact with the maternal blood. The labyrinth layer of the Nrk KO conceptus was gigantic, with enlarged cytoplasm and Golgi bodies in the TGCs. To investigate the function of Nrk in the labyrinth layer, a differentially expressed gene (DEG) analysis was performed. The DEG analysis revealed that labor-promoting factors, such as prostaglandins, were decreased, and pregnancy-maintaining factors, such as the prolactin family and P4 receptor, were increased. These findings suggest that the Nrk KO mice exhibit delayed delivery owing to high P4 concentrations caused by the hypersecretion of pregnancy-maintaining factors, such as PL-2, from the placenta.