Merck
  • Exercise increases Rho-kinase activity and insulin signaling in skeletal muscle.
Exercise increases Rho-kinase activity and insulin signaling in skeletal muscle.
Journal of cellular physiology (2017-12-09)
Vitor R Muñoz, Rafael C Gaspar, Gabriel K Kuga, Alisson L da Rocha, Barbara M Crisol, José D Botezelli, Igor L Baptista, Rania A Mekary, Adelino S R da Silva, Dennys E Cintra, Leandro P de Moura, Eduardo R Ropelle, José R Pauli
ABSTRACT
The effects of physical exercise on insulin signaling and glycemic homeostasis are not yet fully understood. Recent findings elucidated the positive role of Rho-kinase (Rock) in increasing the glucose uptake through insulin receptor substrate-1 (IRS1) phosphorylation in the skeletal muscle. Here, we explored the effects of short-term exercise on Rock activity and insulin signaling. Fischer 344 rats (3 months old) were subjected to a short-term swimming exercise for 2 hr per day for 5 days, with an overload corresponding to 1.5% of body weight. As expected, the exercised group had a reduced glycemia and increased insulin sensitivity. The contents of Rock1, Rock2, and Rock activity were improved in the skeletal muscle of the exercised rats. The contents of RhoA and RhoGEF, which are proteins involved in the Rock metabolism, were also increased in the skeletal muscle after exercise. These changes in the protein contents were accompanied by an increase in the insulin signaling pathway (pIRS1/pPDK/pAkt/pGSK3β/pAS160/GLUT4), Rock activity, and IRS1 phosphorylation at the 632/635 serine residues. On the other hand, when Rock was inhibited with the Y-27632, the insulin sensitivity in response to exercise was impaired. Based on these findings, we conclude that the short-term exercise increased both insulin sensitivity and glucose tolerance, through the increased Rock activity and pIRS1 (serine 632/635) mediated by Rock, in the skeletal muscle of Fischer 344 rats. These data represent an exercise-mediated novel mechanism, suggesting an essential role of Rock activity in the insulin signaling and glucose homeostasis improvement.
Sigma-Aldrich
Anti-phospho-IR (Tyr972) Antibody, Upstate®, from rabbit