- Triphlorethol-A attenuates U251 human glioma cancer cell proliferation and ameliorates apoptosis through JAK2/STAT3 and p38 MAPK/ERK signaling pathways.
Triphlorethol-A attenuates U251 human glioma cancer cell proliferation and ameliorates apoptosis through JAK2/STAT3 and p38 MAPK/ERK signaling pathways.
Glioma is the foremost recurrent type of brain tumor in humans; in particular, glioblastoma (GBM) is the main form of brain tumor (GBM) that is highly proliferative and impervious to apoptosis. Triphlorethol-A (TA), a phlorotannin isolated from Ecklonia cava, exhibited cytoprotective, antioxidant, and anticancer properties. However, the exact molecular action of TA in the U251 human GBM cells remains unknown. This may be the first report on the antiproliferative and apoptotic mechanisms of TA on GBM. The cytotoxicity, intracellular reactive oxygen species (ROS), matrix metalloproteinase (MMP), and cell apoptosis activity of TA have been evaluated by the MTT assay and by DCFH-DA, Rh-123, AO/EB, and western blot analysis. The results obtained showed that TA abridged the viability of U251 cells, while MMP increased apoptosis by increasing the ROS levels in a time-dependent manner. The results showed that a reduction in U251 cell proliferation was associated with the regulation of JAK2/STAT3 and p38 MAPK/ERK signaling pathways. TA was found to suppress pJAK, pSTAT3, p38 MAPK, and pERK phosphorylation, thereby causing Bax/Bcl-2 imbalance, activating the caspase cascade and cytochrome c, and inducing apoptosis. Our findings showed that the suppression of JAK2/STAT3 and p38 MAPK/ERK signaling by TA results in cell growth arrest and stimulation of apoptosis in GBM cells. These studies justify the protective remedy of TA against GBM.