Merck
  • A Flexible Solid Composite Electrolyte with Vertically Aligned and Connected Ion-Conducting Nanoparticles for Lithium Batteries.
A Flexible Solid Composite Electrolyte with Vertically Aligned and Connected Ion-Conducting Nanoparticles for Lithium Batteries.
Nano letters (2017-04-15)
Haowei Zhai, Pengyu Xu, Mingqiang Ning, Qian Cheng, Jyotirmoy Mandal, Yuan Yang
ABSTRACT
Replacing flammable organic liquid electrolytes with solid Li-ion conductors is a promising approach to realize safe rechargeable batteries with high energy density. Composite solid electrolytes, which are comprised of a polymer matrix with ceramic Li-ion conductors dispersed inside, are attractive, since they combine the flexibility of polymer electrolytes and high ionic conductivities of ceramic electrolytes. However, the high conductivity of ceramic fillers is largely compromised by the low conductivity of the matrix, especially when nanoparticles (NPs) are used. Therefore, optimizations of the geometry of ceramic fillers are critical to further enhance the conductivity of composite electrolytes. Here we report the vertically aligned and connected Li1+xAlxTi2-x(PO4)3 (LATP) NPs in the poly(ethylene oxide) (PEO) matrix to maximize the ionic conduction, while maintaining the flexibility of the composite. This vertically aligned structure can be fabricated by an ice-templating-based method, and its conductivity reaches 0.52 × 10-4 S/cm, which is 3.6 times that of the composite electrolyte with randomly dispersed LATP NPs. The composite electrolyte also shows enhanced thermal and electrochemical stability compared to the pure PEO electrolyte. This method opens a new approach to optimize ion conduction in composite solid electrolytes for next-generation rechargeable batteries.
Sigma-Aldrich
Lithium perchlorate, anhydrous, ≥99.9% trace metals basis