Merck
Cerebral Ischemic Preconditioning Aggravates Death of Oligodendrocytes.
Biomolecules (2022-12-24)
Teng Guan, Ying Guo, Chengren Li, Ting Zhou, Qiang Yu, Chaoxian Yang, Guohui Zhang, Jiming Kong
ABSTRACT
Neurodegeneration can benefit from ischemic preconditioning, a natural adaptive reaction to sublethal noxious stimuli. Although there is growing interest in advancing preconditioning to preserve brain function, preconditioning is not yet considered readily achievable in clinical settings. One of the most challenging issues is that there is no fine line between preconditioning stimuli and lethal stimuli. Here, we show deleterious effect of preconditioning on oligodendrocyte precursor cells (OPCs). We identified Bcl-2/adenovirus E1B 19-kDa interacting protein 3 (BNIP3), a mitochondrial BH3-only protein specifically involved in OPCs loss after preconditioning. Repeated ischemia stabilized BNIP3 and increased the vulnerability of OPCs to subsequent ischemic events. BNIP3 became mitochondrial-bound and was concurrent with the dysfunction of monocarboxylate transporter 1 (MCT1). Inhibition of BNIP3 by RNAi or necrostatin-1 (Nec-1) and knocking out of BNIP3 almost completely prevented OPCs loss and preserved white matter integrity. Together, our results suggest that the unfavorable effect of BNIP3 on OPCs should be noted for safe development of ischemic tolerance. BNIP3 inhibition appears to be a complementary approach to improve the efficacy of preconditioning for ischemic stroke.
Sigma-Aldrich
Anti-NG2 Antibody, clone 132.38, clone 132.38, Upstate®, from mouse
Sigma-Aldrich
Anti-A2B5 Antibody, clone A2B5-105, clone A2B5-105, Chemicon®, from mouse