Merck
  • Production of Class II MHC Proteins in Lentiviral Vector-Transduced HEK-293T Cells for Tetramer Staining Reagents.
Production of Class II MHC Proteins in Lentiviral Vector-Transduced HEK-293T Cells for Tetramer Staining Reagents.
Current protocols (2021-02-05)
Richard A Willis, Vasanthi Ramachandiran, John C Shires, Ge Bai, Kelly Jeter, Donielle L Bell, Lixia Han, Tamara Kazarian, Kyla C Ugwu, Oskar Laur, Susana Contreras-Alcantara, Dale L Long, John D Altman
ABSTRACT
Class II major histocompatibility complex peptide (MHC-IIp) multimers are precisely engineered reagents used to detect T cells specific for antigens from pathogens, tumors, and self-proteins. While the related Class I MHC/peptide (MHC-Ip) multimers are usually produced from subunits expressed in E. coli, most Class II MHC alleles cannot be produced in bacteria, and this has contributed to the perception that MHC-IIp reagents are harder to produce. Herein, we present a robust constitutive expression system for soluble biotinylated MHC-IIp proteins that uses stable lentiviral vector-transduced derivatives of HEK-293T cells. The expression design includes allele-specific peptide ligands tethered to the amino-terminus of the MHC-II β chain via a protease-cleavable linker. Following cleavage of the linker, HLA-DM is used to catalyze efficient peptide exchange, enabling high-throughput production of many distinct MHC-IIp complexes from a single production cell line. Peptide exchange is monitored using either of two label-free methods, native isoelectric focusing gel electrophoresis or matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry of eluted peptides. Together, these methods produce MHC-IIp complexes that are highly homogeneous and that form the basis for excellent MHC-IIp multimer reagents. © 2021 Wiley Periodicals LLC. Basic Protocol 1: Lentivirus production and expression line creation Support Protocol 1: Six-well assay for estimation of production cell line yield Support Protocol 2: Universal ELISA for quantifying proteins with fused leucine zippers and His-tags Basic Protocol 2: Cultures for production of Class II MHC proteins Basic Protocol 3: Purification of Class II MHC proteins by anti-leucine zipper affinity chromatography Alternate Protocol 1: IMAC purification of His-tagged Class II MHC Support Protocol 3: Protein concentration measurements and adjustments Support Protocol 4: Polishing purification by anion-exchange chromatography Support Protocol 5: Estimating biotinylation percentage by streptavidin precipitation Basic Protocol 4: Peptide exchange Basic Protocol 5: Analysis of peptide exchange by matrix-assisted laser desorption/ionization (MALDI) mass spectrometry Alternate Protocol 2: Native isoelectric focusing to validate MHC-II peptide loading Basic Protocol 6: Multimerization Basic Protocol 7: Staining cells with Class II MHC tetramers.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)