Merck
  • Grade-dependent changes in sphingolipid metabolism in clear cell renal cell carcinoma.
Grade-dependent changes in sphingolipid metabolism in clear cell renal cell carcinoma.
Journal of cellular biochemistry (2022-02-22)
Grzegorz Młynarczyk, Agnieszka Mikłosz, Jarosław Suchański, Safoura Reza, Lech Romanowicz, Krzysztof Sobolewski, Adrian Chabowski, Marcin Baranowski
ABSTRACT
There is a host of evidence for the role of bioactive sphingolipids in cancer biology, and dysregulated sphingolipid metabolism was observed in many malignant tumors. The aim of the present study was to provide more detailed data on sphingolipid metabolism in different stages of clear cell renal cell carcinoma (ccRCC). Samples of the tumor and noncancerous fragments of the same kidney were collected from patients who underwent a radical nephrectomy. The subjects were stratified according to the degree of malignancy of the tumor (n = 14 for G2, 12 for G3, and 9 for G4). The content of bioactive sphingolipids/glycosphingolipids was measured with an HPLC and HPTLC method, and the mRNA and protein expression of sphingolipid transporters and metabolizing enzymes was evaluated using real-time polymerase chain reaction (PCR) and Western blot, respectively. Compared to healthy kidney tissue, ccRCC was characterized by accumulation of sphingosine, sphingosine-1-phosphate (S1P), ceramide, dihydrosphingosine, and dihydroceramide. However, in the case of the latter two, the accumulation was limited to higher malignancy grades. In addition, compared to the healthy tissue, the content of gangliosides in the tumor was increased at the expense of globosides. We also found profound grade-dependent changes in the mRNA level of S1P-metabolizing enzymes, and spinster homolog 2. In general, their expression was much higher in G2 tumors compared to higher malignancy grades. We conclude that ccRCC is characterized by profound and multilevel alterations in sphingolipid metabolism, which to a large extent are grade-dependent. We hypothesize that dysregulation of sphingolipid metabolism contributes to the progression of ccRCC.
Sigma-Aldrich
Anti-SPNS2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-SGPL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution