Merck
  • Fermented Jussara: Evaluation of Nanostructure Formation, Bioaccessibility, and Antioxidant Activity.
Fermented Jussara: Evaluation of Nanostructure Formation, Bioaccessibility, and Antioxidant Activity.
Frontiers in bioengineering and biotechnology (2022-04-01)
Michele Amendoeira Giaconia, Sergiana Dos Passos Ramos, Camilly Fratelli, Marcelo Assis, Tatiana Martelli Mazzo, Elson Longo, Veridiana Vera de Rosso, Anna Rafaela Cavalcante Braga
ABSTRACT
Among the species of plants present in the Atlantic Forest, the jussara (Euterpe edulis Mart.) stands out for the contents of bioactive compounds present in its composition. Fermentation processes can be essential in converting bioproducts and bioactive compounds, improving their biological properties. In addition, the improvement of procedures for the maintenance of the features of bioactive compounds has been a research focus in recent years, and the nanotechnology features that can potentially solve this issue have been highlighted among the most reviewed paths. The present work focused on tailoring nanostructures applying polyethylene oxide, assembling fermented jussara pulp nanofibers, and assessing their characteristics. The results revealed the formation of fermented jussara nanofibers with a diameter of 101.2 ± 26.2 nm. Also, the obtained results allow us to state that it is possible to maintain or even increase the antioxidant activity of anthocyanins and their metabolites after fermentation processes.
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications